Assay Details
| Gene Symbol: Zfp799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293229.1 | NP_001280158.1 | 2 - 3 | 301 | 101 | |
| GenBank mRNA | BC062813.1 | - | 2 - 3 | 255 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 811 | Zfp811 | BC052046 | -11.5kb | Sense | Chr. 17 - 32795676 - 32809931 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 870 | Zfp870 | AI173952 | 57.0kb | Sense | Chr. 17 - 32879221 - 32891603 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Zfp799
Genomic Map