Assay Details
| Gene Symbol: LOC545966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC079901.1 | - | 1 - 2 | 259 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | E2F transcription factor 8 | E2f8 |
4432406C08Rik, AA410048 |
-16.7kb | Anti-sense | Chr. 7 - 48866429 - 48882142 on NCBI Build GRCm38 |
| Downstream | neuron navigator 2 | Nav2 |
5330421F07Rik, AU067780, BQ960892, HELAD1, POMFIL2, RAINB1, RAINB2, Unc53H2, mKIAA3015 |
51.1kb | Sense | Chr. 7 - 48959066 - 49610088 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

LOC545966
Genomic Map