Assay Details
| Gene Symbol: Gm5782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC079904.1 | - | 1 - 2 | 228 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 146 | Mir146 |
Mirn146, miR-146a, mmu-mir-146 |
-41.7kb | Sense | Chr. 11 - 43374397 - 43374461 on NCBI Build GRCm38 |
| Downstream | pituitary tumor-transforming gene 1 | Pttg1 |
AW555095, C87862, Pttg, Pttg3 |
3.4kb | Sense | Chr. 11 - 43420248 - 43426248 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm5782
Genomic Map