Assay Details
| Gene Symbol: Pantr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC094545.1 | - | 327 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930448I06 gene | 4930448I06Rik | -1.4mb | Sense | Chr. 1 - 41167560 - 41181252 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Pantr1
Genomic Map