Assay Details
Gene Symbol: Gm2511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC100510.1 | - | 2 - 3 | 165 | 82 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | CN836674.1 | - | 168 | Chr. - - [-] on Build | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4931406B18 gene | 4931406B18Rik | 4930538L19Rik | -8.7kb | Sense | Chr. 7 - 43492044 - 43505969 on NCBI Build GRCm38 |
Downstream | CD33 antigen | Cd33 |
Siglec-3, gp67 |
11.7kb | Sense | Chr. 7 - 43527399 - 43544428 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
