Assay Details
| Gene Symbol: Hist1h3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M15434.1 | - | 1 | 46 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
histone cluster 1, H2af |
Hist1h2af | H2a-221 | -1.5kb | Sense | Chr. 13 - 23533911 - 23534378 on NCBI Build GRCm38 |
| Downstream |
histone cluster 1, H2bh |
Hist1h2bh | H2b-221 | 6.6kb | Anti-sense | Chr. 13 - 23542923 - 23543444 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hist1h3g
Genomic Map