Assay Details
| Gene Symbol: Olfr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146921.2 | NP_667132.1 | 2 - 3 | 288 | 95 | |
| GenBank mRNA | DR065653.1 | - | 2 - 3 | 288 | 95 | |
| S73846.1 | - | 2 - 3 | 236 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | olfactory receptor 376 | Olfr376 | MOR135-12 | -20.3kb | Anti-sense | Chr. 11 - 73371246 - 73375704 on NCBI Build GRCm38 |
| Downstream |
olfactory receptor 75, pseudogene 1 |
Olfr75-ps1 |
MOR135-20, Olfr75, V1 |
8.7kb | Sense | Chr. 11 - 73405450 - 73409858 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_146921.2, S73846.1 transcripts.
Set Membership: |
|

Olfr1
Genomic Map