Assay Details
| Gene Symbol: 2310061L18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK010009.1 | - | 1 | 552 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | frizzled homolog 7 (Drosophila) | Fzd7 | Fz7 | -28.8kb | Anti-sense | Chr. 1 - 59482147 - 59486955 on NCBI Build GRCm38 |
| Downstream | SMT3 suppressor of mif two 3 homolog 1 (yeast) | Sumo1 |
GMP1, PIC1, SENTRIN, SMT3, SMT3H3, SMTP3, SUMO-1, Smt3C, Ubl1 |
122.8kb | Sense | Chr. 1 - 59639434 - 59670834 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2310061L18Rik
Genomic Map