Assay Details
| Gene Symbol: 2310068C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK010104.1 | - | 1 | 1152 | 70 | |
| BG061907.2 | - | 1 | 70 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | paraneoplastic antigen MA2 | Pnma2 |
A830049P17Rik, mKIAA0883 |
-71.1kb | Anti-sense | Chr. 14 - 66900682 - 66920063 on NCBI Build GRCm38 |
| Downstream |
protein phosphatase 2, regulatory subunit B, alpha |
Ppp2r2a |
2410004D02Rik, mKIAA1541 |
21.6kb | Sense | Chr. 14 - 67014056 - 67072471 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2310068C19Rik
Genomic Map