Assay Details
| Gene Symbol: 2410049M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK010698.1 | - | 1 | 247 | 118 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ariadne homolog 2 (Drosophila) | Arih2 |
AI843547, ARI2, TRIAD1, UIP48 |
Sense | Chr. 9 - 108602942 - 108654719 on NCBI Build GRCm38 | |
| Downstream |
solute carrier family 25 (mitochondrial carnitine/acylcarnitine translocase), member 20 |
Slc25a20 |
1110007P09Rik, C78826, Cact, mCAC |
7.4kb | Anti-sense | Chr. 9 - 108662098 - 108684641 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2410049M19Rik
Genomic Map