Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
277656 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798407 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898825 |
XM_001477378.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
898825 |
XM_001480808.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
478930 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
872440 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
461792 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
650804 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
221375 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
23677 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
7181 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135909 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P102280 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135909 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00965631 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P102280 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P103179 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P102280 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135909 |
- |
- |