Assay Details
| Gene Symbol: 2600002B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA727298.1 | - | 1 | 109 | 72 | |
| AI586157.1 | - | 1 | 72 | |||
| AK011142.1 | - | 1 | 1050 | 72 | ||
| BI465735.1 | - | 1 | 72 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
zinc finger, CCHC domain containing 14 |
Zcchc14 |
AA792890, BDG29 |
Sense | Chr. 8 - 121598697 - 121652761 on NCBI Build GRCm38 | |
| Downstream | junctophilin 3 | Jph3 |
JP-3, Jp3 |
112.7kb | Anti-sense | Chr. 8 - 121730559 - 121791083 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2600002B07Rik
Genomic Map