Assay Details
| Gene Symbol: 2610005G05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011318.1 | - | 1 | 550 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | geminin coiled-coil domain containing | Gmnc |
Gemc1, Gm1778, Gm606 |
-228.0 | Anti-sense | Chr. 16 - 26957235 - 26991667 on NCBI Build GRCm38 |
| Downstream | osteocrin | Ostn | Ostc | 314.4kb | Sense | Chr. 16 - 27306983 - 27351209 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2610005G05Rik
Genomic Map