Assay Details
| Gene Symbol: 2610007B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011331.1 | - | 1 | 1750 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6812 | Gm6812 | EG627927 | -516.2kb | Anti-sense | Chr. X - 68892373 - 68893053 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700111N16 gene | 1700111N16Rik | 518.1kb | Anti-sense | Chr. X - 69929260 - 70143588 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2610007B07Rik
Genomic Map