Assay Details
| Gene Symbol: 2610011I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011377.1 | - | 1 | 1352 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC048403 | BC048403 | E030027L10 | -273.7kb | Anti-sense | Chr. 10 - 121739603 - 121757613 on NCBI Build GRCm38 |
| Downstream | transmembrane emp24 domain trafficking protein 2 pseudogene | Gm9079 | 45.3kb | Sense | Chr. 10 - 122078112 - 122079879 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2610011I18Rik
Genomic Map