Assay Details
| Gene Symbol: 2610037P13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI852391.1 | - | 1 | 96 | ||
| AK011718.1 | - | 1 | 557 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 955B | Zfp955b |
A430003O12Rik, C430039G02Rik, C530035A18Rik, Gm4455, Zfp883b |
-24.7kb | Anti-sense | Chr. 17 - 33289517 - 33304689 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 101 | Zfp101 |
AI316498, AI325003 |
50.0kb | Sense | Chr. 17 - 33380174 - 33394637 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2610037P13Rik
Genomic Map