Assay Details
| Gene Symbol: 2610303G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011970.1 | - | 1 - 2 | 222 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calsyntenin 2 | Clstn2 |
2900042C18Rik, AI448973, CSTN2, Cs2, Cst-2, mKIAA4134 |
-153.1kb | Sense | Chr. 9 - 97444395 - 98033384 on NCBI Build GRCm38 |
| Downstream | nicotinamide nucleotide adenylyltransferase 3 | Nmnat3 |
4933408N02Rik, PNAT3 |
100.3kb | Anti-sense | Chr. 9 - 98287432 - 98420442 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

2610303G11Rik
Genomic Map