Assay Details
| Gene Symbol: 2700005E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA146492.1 | - | 1 | 392 | 73 | |
| AK012210.1 | - | 1 | 155 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dopachrome tautomerase | Dct |
DT, TRP-2, TRP2, Tyrp-2, Tyrp2, slaty, slt |
-39.0kb | Anti-sense | Chr. 14 - 118012790 - 118052323 on NCBI Build GRCm38 |
| Downstream | TDP-glucose 4,6-dehydratase | Tgds |
2610017J16Rik, 2610025M23Rik, AI648925 |
20.1kb | Anti-sense | Chr. 14 - 118111911 - 118132765 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2700005E23Rik
Genomic Map