Assay Details
| Gene Symbol: 2700022O18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012274.1 | - | 1 | 845 | 83 | |
| AW488528.1 | - | 1 | 83 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
poly (ADP-ribose) polymerase family, member 4 |
Parp4 |
Adprtl1, C030027K23Rik, E230037B21Rik, Gm743, PARPL, PH5P, VAULT3, VPARP, p193 |
-7.3kb | Anti-sense | Chr. 14 - 56575619 - 56659798 on NCBI Build GRCm38 |
| Downstream | M-phase phosphoprotein 8 | Mphosph8 |
1500035L22Rik, 4930548G07Rik, AU040635 |
126.0 | Anti-sense | Chr. 14 - 56668240 - 56697430 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2700022O18Rik
Genomic Map