| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
833032 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
620482 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
593257 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
69647 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
358450 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
708830 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
641363 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
339604 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
558728 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
587877 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
842760 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693507 |
AF109136.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693507 |
AK051676.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693507 |
AK078842.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693507 |
AK089898.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693507 |
AK156015.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693507 |
AK156517.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693507 |
BC068313.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693507 |
NM_010576.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693507 |
U97151.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
693507 |
X53176.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
688307 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
824675 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
429869 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
217327 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
122929 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P130550 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02353702 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919905 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02505983 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919904 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02792176 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919822 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919916 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919935 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03074279 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919940 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02717170 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919928 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919950 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919949 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919908 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919930 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02717174 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03006007 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02353705 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919831 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919912 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919830 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919943 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919913 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03059017 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919914 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919945 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919931 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919946 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919933 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919826 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919823 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919929 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02557366 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02717173 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919942 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03074291 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919820 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919951 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21919932 |
- |
- |