Assay Details
| Gene Symbol: 2700089I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045308.1 | 2 - 3 | 208 | 72 | ||
| GenBank mRNA | AK012591.1 | - | 2 - 3 | 209 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | empty spiracles homeobox 2 | Emx2 | Pdo | -28.2kb | Sense | Chr. 19 - 59458116 - 59465357 on NCBI Build GRCm38 |
| Downstream | RAB11 family interacting protein 2 (class I) | Rab11fip2 |
4930470G04Rik, A830046J09Rik, AW558126, Nrip11 |
204.0kb | Anti-sense | Chr. 19 - 59760441 - 59943666 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

2700089I24Rik
Genomic Map