Assay Details
| Gene Symbol: 2700089I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045308.1 | 4 - 5 | 455 | 113 | ||
| GenBank mRNA | AK012591.1 | - | 4 - 5 | 456 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | empty spiracles homeobox 2 | Emx2 | Pdo | -92.1kb | Sense | Chr. 19 - 59458116 - 59465357 on NCBI Build GRCm38 |
| Downstream | RAB11 family interacting protein 2 (class I) | Rab11fip2 |
4930470G04Rik, A830046J09Rik, AW558126, Nrip11 |
201.1kb | Anti-sense | Chr. 19 - 59760441 - 59943666 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2215404 | AK012591.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 64855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724896 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647430 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342646 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 585161 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162384 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740716 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935735 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319398 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801940 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852451 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313765 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615684 | XM_890986.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615684 | XM_907796.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528433 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626970 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291173 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266773 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 28825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163730 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36086 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133204 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03630127 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09217676 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133204 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09217626 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09217614 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102709 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09217626 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14328987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14385704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14483077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14254072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14507002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14257580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14343884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14385707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14257579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219775 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14390553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14469253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14215274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32219777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14329352 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14483078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14343937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14215277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14257592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14254073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32220151 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1431912_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431912_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431912_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900007403.mm.1 | NR_045308.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007403.mm.1 | NR_045308.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900007403.mm.1 | TC1900000856.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431912_at | - | - |
| Mouse Expression Set 430 | 1431912_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431912_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431912_at | - | - |

2700089I24Rik
Genomic Map