Assay Details
| Gene Symbol: 2810048H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012924.1 | - | 1 | 746 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 72 | Tmem72 | C230095G01Rik | -427.3kb | Sense | Chr. 6 - 116679103 - 116716978 on NCBI Build GRCm38 |
| Downstream | chemokine (C-X-C motif) ligand 12 | Cxcl12 |
Pbsf, Scyb12, Sdf1, Tlsf, Tpar1 |
23.4kb | Anti-sense | Chr. 6 - 117168535 - 117181368 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2810048H02Rik
Genomic Map