Assay Details
| Gene Symbol: Tbx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023814.4 | NP_076303.3 | 8 | 4453 | 100 | |
| GenBank mRNA | AK012980.1 | - | 1 | 1553 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | centrosomal protein 162 | Cep162 |
4922501C03Rik, mKIAA1009 |
-447.3kb | Sense | Chr. 9 - 87189570 - 87255536 on NCBI Build GRCm38 |
| Downstream | predicted gene 28231 | Gm28231 | 555.3kb | Anti-sense | Chr. 9 - 88259887 - 88263035 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tbx18
Genomic Map