Assay Details
| Gene Symbol: 2810404I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012984.1 | - | 1 | 1851 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | R3H domain and coiled-coil containing 1 like | R3hcc1l |
1700036B12Rik, AI450607, C10orf28, D19Ertd386e, GIDRP86, Gidrp88 |
Sense | Chr. 19 - 42518312 - 42592256 on NCBI Build GRCm38 | |
| Downstream | lysyl oxidase-like 4 | Loxl4 |
4833426I20Rik, Loxc |
24.3kb | Anti-sense | Chr. 19 - 42592279 - 42612906 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2810404I24Rik
Genomic Map