Assay Details
| Gene Symbol: 2810414N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013093.1 | - | 1 | 252 | 121 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 129, member A |
Fam129a |
AI256368, AU019833, Niban |
-27.0kb | Anti-sense | Chr. 1 - 151571373 - 151721939 on NCBI Build GRCm38 |
| Downstream |
ER degradation enhancer, mannosidase alpha-like 3 |
Edem3 | 2310050N11Rik | 5.3kb | Anti-sense | Chr. 1 - 151755344 - 151822328 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2810414N06Rik
Genomic Map