Assay Details
| Gene Symbol: 2810439L12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013270.1 | - | 1 | 252 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
autophagy related 4C, cysteine peptidase |
Atg4c |
Apg4-C, Apg4c, Atg4cl, Autl1 |
-358.5kb | Anti-sense | Chr. 4 - 99190891 - 99297258 on NCBI Build GRCm38 |
| Downstream | forkhead box D3 | Foxd3 |
CWH3, Genesis, Hfh2 |
286.0 | Anti-sense | Chr. 4 - 99656299 - 99658671 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2810439L12Rik
Genomic Map