Assay Details
| Gene Symbol: 2810455D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013353.1 | - | 1 | 1153 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein prenyltransferase alpha subunit repeat containing 1 | Ptar1 |
1700084D09Rik, 2010013L17Rik, 4930428J16Rik, AI449677 |
-6.9kb | Sense | Chr. 19 - 23686898 - 23723092 on NCBI Build GRCm38 |
| Downstream |
amyloid beta (A4) precursor protein binding, family A, member 1 |
Apba1 |
6430513E09Rik, Lin-10, Mint, Mint1, Urop11, X11, X11alpha |
27.2kb | Sense | Chr. 19 - 23758876 - 23949598 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2810455D13Rik
Genomic Map