Assay Details
| Gene Symbol: Ildr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164528.1 | NP_001158000.1 | 10 | 6778 | 81 | |
| GenBank mRNA | AK013417.1 | - | 1 | 249 | 81 | |
| BC035277.1 | - | 1 | 2702 | 81 | ||
| G49232.1 | - | 1 | 81 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DNA segment, Chr 16, ERATO Doi 472, expressed |
D16Ertd472e |
1700010I10Rik, 2310009O17Rik, E330003K22Rik, Eurl |
Anti-sense | Chr. 16 - 78540336 - 78576688 on NCBI Build GRCm38 | |
| Downstream | transcriptional adaptor 1 | Tada1 |
2900026B15Rik, D1Ertd251e, Staf42, Tada1l |
63.7kb | Sense | Chr. 1 - 166379167 - 166393621 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ildr2
Genomic Map