Assay Details
| Gene Symbol: 2900001G08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013461.1 | - | 1 | 1253 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | glucagon-like peptide 1 receptor | Glp1r |
GLP-1R, GLP1Rc |
-2.4kb | Sense | Chr. 17 - 30901867 - 30936510 on NCBI Build GRCm38 |
| Downstream | uromodulin-like 1 | Umodl1 | D17Ertd488e | 13.9kb | Sense | Chr. 17 - 30954640 - 31010710 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900001G08Rik
Genomic Map