Assay Details
| Gene Symbol: 2900006G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013492.1 | - | 1 | 155 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 29907 |
Gm29907 | -957.7kb | Sense | Chr. 16 - 39984595 - 40264481 on NCBI Build GRCm38 | |
| Downstream | limbic system-associated membrane protein | Lsamp |
5430428I19, AW046674, B130007O04Rik, D930023J12Rik, Lam, Lamp |
310.2kb | Sense | Chr. 16 - 41532788 - 42181679 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900006G07Rik
Genomic Map