Assay Details
| Gene Symbol: 2900012M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013530.1 | - | 1 | 1354 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6237 | Mir6237 |
Gm27411, mmu-mir-6237 |
-894.6kb | Anti-sense | Chr. 9 - 9894431 - 9894521 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930568E12 gene | 4930568E12Rik | 2.3mb | Sense | Chr. 9 - 13091267 - 13105855 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900012M01Rik
Genomic Map