Assay Details
| Gene Symbol: 2900034C19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013612.1 | - | 1 | 1153 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 12718 | Gm12718 | OTTMUSG00000008109 | -219.5kb | Sense | Chr. 4 - 103382500 - 103492188 on NCBI Build GRCm38 |
| Downstream |
complement component 8, beta polypeptide |
C8b |
4930439B20Rik, AI595927 |
1.1mb | Sense | Chr. 4 - 104764576 - 104804549 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900034C19Rik
Genomic Map