Assay Details
| Gene Symbol: Kantr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013706.1 | - | 1 | 452 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lysine (K)-specific demethylase 5C | Kdm5c |
D930009K15Rik, Jarid1c, Smcx, mKIAA0234 |
-36.1kb | Anti-sense | Chr. X - 152233196 - 152279102 on NCBI Build GRCm38 |
| Downstream | TSPY-like 2 | Tspyl2 |
CDA1, CINAP, DENTT, DXBwg1396e, DXHXS1008E, E130307F10Rik |
21.1kb | Sense | Chr. X - 152336852 - 152342506 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kantr
Genomic Map