Assay Details
| Gene Symbol: 2900074G08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013781.1 | - | 1 | 449 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synaptotagmin I | Syt1 |
AW124717, G630098F17Rik, SytI |
Sense | Chr. 10 - 108497650 - 109010975 on NCBI Build GRCm38 | |
| Downstream | neuron navigator 3 | Nav3 |
4732483H20Rik, 9630020C08Rik, POMFIL1, Pomfil1p, mKIAA0938, unc53H3 |
907.0kb | Sense | Chr. 10 - 109681573 - 110000391 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900074G08Rik
Genomic Map