Assay Details
| Gene Symbol: 2900076A13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013795.1 | - | 1 | 455 | 103 | |
| BG075471.2 | - | 1 | 103 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA F830045P16 gene | F830045P16Rik | Sirpb3 | -59.7kb | Anti-sense | Chr. 2 - 129457797 - 129536602 on NCBI Build GRCm38 |
| Downstream | prodynorphin | Pdyn | Dyn | 89.3kb | Anti-sense | Chr. 2 - 129686549 - 129699938 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900076A13Rik
Genomic Map