Assay Details
| Gene Symbol: 2900086E13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013824.1 | - | 1 | 845 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5 | St6galnac5 |
AI851940, Siat7e |
Sense | Chr. 3 - 152820709 - 152982207 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930482G09 gene | 4930482G09Rik | 181.5kb | Sense | Chr. 3 - 153147943 - 153152108 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900086E13Rik
Genomic Map