Assay Details
| Gene Symbol: 3100002H20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK013917.1 | - | 1 | 450 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
potassium channel, subfamily K, member 9 |
Kcnk9 | Task3 | -237.9kb | Sense | Chr. 15 - 72502927 - 72547853 on NCBI Build GRCm38 |
| Downstream | paternally expressed 13 | Peg13 | 18.5kb | Sense | Chr. 15 - 72805604 - 72810324 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

3100002H20Rik
Genomic Map