Assay Details
| Gene Symbol: 3110005L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA611589.1 | - | 1 | 372 | 91 | |
| AK014001.1 | - | 1 | 1048 | 91 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dihydrolipoamide S-acetyltransferase (E2 component of pyruvate dehydrogenase complex) | Dlat |
6332404G05Rik, DLTA, PDC-E2 |
-749.0 | Anti-sense | Chr. 9 - 50634633 - 50659780 on NCBI Build GRCm38 |
| Downstream | DIX domain containing 1 | Dixdc1 |
4930563F16Rik, BC048182, Ccd1, Ccd1Aalpha1L, Ccd1Abeta1L, Ccd1BalphaL, Ccd1BbetaL, Ccd1CL, mKIAA1735 |
823.0 | Anti-sense | Chr. 9 - 50662752 - 50737737 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

3110005L24Rik
Genomic Map