Assay Details
| Gene Symbol: Fam184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014044.1 | - | 1 | 351 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | minichromosome maintenance complex component 9 | Mcm9 |
9030408O17Rik, Gm235, Mcmdc1 |
-2.7kb | Sense | Chr. 10 - 53536315 - 53630439 on NCBI Build GRCm38 |
| Downstream |
mannosidase 1, alpha |
Man1a |
Man1a1, PCR1 |
270.5kb | Sense | Chr. 10 - 53904785 - 54076586 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam184a
Genomic Map