Assay Details
| Gene Symbol: 3110045A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014176.1 | - | 1 | 1145 | 92 | |
| AV166410.1 | - | 1 | 122 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102634389 | LOC102634389 | -121.0kb | Anti-sense | Chr. 15 - 95824458 - 95830072 on NCBI Build GRCm38 | |
| Downstream | nuclear receptor coactivator 2 pseudogene | D030018L15Rik | 99.2kb | Anti-sense | Chr. 15 - 96051616 - 96079553 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

3110045A19Rik
Genomic Map