Assay Details
Gene Symbol: Zfp942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014195.1 | - | 1 | 1446 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 2210404O09 gene | 2210404O09Rik | -20.2kb | Sense | Chr. 17 - 21878907 - 21909926 on NCBI Build GRCm38 | |
Downstream | zinc finger prtoein 943 | Zfp943 | 4930432O21Rik | 30.1kb | Anti-sense | Chr. 17 - 21962559 - 21994366 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
