Assay Details
| Gene Symbol: 3732407C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014404.1 | - | 1 | 3350 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 34637 |
Gm34637 | -52.9kb | Anti-sense | Chr. 12 - 12667732 - 12669963 on NCBI Build GRCm38 | |
| Downstream |
v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) |
Mycn |
N-myc, Nmyc, Nmyc-1, Nmyc1, bHLHe37, c-nmyc |
209.7kb | Anti-sense | Chr. 12 - 12936093 - 12941836 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

3732407C23Rik
Genomic Map