Assay Details
| Gene Symbol: Fam160a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242363.2 | NP_001229292.1 | 13 | 5657 | 82 | |
| NM_001242364.2 | NP_001229293.1 | 12 | 5608 | 82 | ||
| NM_001242365.2 | NP_001229294.1 | 12 | 5014 | 82 | ||
| NM_199009.3 | NP_950174.1 | 12 | 5566 | 82 | ||
| GenBank mRNA | AK014591.1 | - | 1 | 2448 | 82 | |
| AK122552.1 | - | 11 | 5315 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 5901 | Gm5901 | EG545989 | Anti-sense | Chr. 7 - 105375080 - 105378377 on NCBI Build GRCm38 | |
| Downstream | cyclic nucleotide gated channel alpha 4 | Cnga4 | 21.4kb | Anti-sense | Chr. 7 - 105400377 - 105409313 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam160a2
Genomic Map