Assay Details
| Gene Symbol: 4733401A01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI642040.1 | - | 1 | 151 | 100 | |
| AK014628.1 | - | 1 | 1257 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/arginine-rich splicing factor 4 | Srsf4 |
5730499P16Rik, AW550192, MNCb-2616, SRp75, Sfrs4 |
Sense | Chr. 4 - 131873611 - 131901725 on NCBI Build GRCm38 | |
| Downstream | predicted gene 12992 | Gm12992 | OTTMUSG00000009742 | 10.1kb | Anti-sense | Chr. 4 - 131899478 - 131920029 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4733401A01Rik
Genomic Map