Assay Details
| Gene Symbol: 4833420L08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014738.1 | - | 1 | 1147 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 6521 | Gm6521 | EG624685 | -17.6kb | Anti-sense | Chr. 4 - 86747532 - 86750859 on NCBI Build GRCm38 |
| Downstream | ribosomal protein S6 | Rps6 | S6R | 84.3kb | Anti-sense | Chr. 4 - 86854099 - 86857367 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4833420L08Rik
Genomic Map