Assay Details
| Gene Symbol: 4921520J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014930.1 | - | 1 | 448 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Tia1 cytotoxic granule-associated RNA binding protein-like 1 | Tial1 |
5330433G13Rik, AL033329, TIAR, mTIAR |
-60.9kb | Sense | Chr. 7 - 128439772 - 128461668 on NCBI Build GRCm38 |
| Downstream | BCL2-associated athanogene 3 | Bag3 |
AA407278, Bis, mg638 |
435.0 | Anti-sense | Chr. 7 - 128523583 - 128546981 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4921520J07Rik
Genomic Map