Assay Details
| Gene Symbol: 4930401B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015030.1 | - | 1 | 952 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 14461 | Gm14461 |
A830093M07, OTTMUSG00000016703 |
-543.3kb | Sense | Chr. 2 - 78237547 - 78302230 on NCBI Build GRCm38 |
| Downstream | ubiquitin-conjugating enzyme E2E 3 | Ube2e3 |
Ubce4, ubcM2 |
21.7kb | Sense | Chr. 2 - 78868243 - 78920720 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930401B11Rik
Genomic Map