Assay Details
| Gene Symbol: 4930402M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015054.1 | - | 1 | 550 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WD repeat domain 76 | Wdr76 |
5830411K18Rik, 6030457D09, AA517836 |
-233.8kb | Sense | Chr. 2 - 121506723 - 121544860 on NCBI Build GRCm38 |
| Downstream | cancer susceptibility candidate 4 | Casc4 |
D130060C09Rik, VGFG2573 |
87.5kb | Sense | Chr. 2 - 121866970 - 121936220 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930402M22Rik
Genomic Map