Assay Details
| Gene Symbol: 4930405A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015090.1 | - | 1 | 554 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
wingless-type MMTV integration site family, member 3 |
Wnt3 |
Int-4, Wnt-3 |
Anti-sense | Chr. 11 - 103774175 - 103818021 on NCBI Build GRCm38 | |
| Downstream | N-ethylmaleimide sensitive fusion protein | Nsf |
AI316878, AU020090, AU067812, SKD2 |
11.8kb | Sense | Chr. 11 - 103821782 - 103954056 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930405A07Rik
Genomic Map